missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals

Product Code. 11372417
Click to view available options
Quantity:
250 mg
1 g
5 g

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-49-5
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00064417
InChI Key BHRZNVHARXXAHW-BYPYZUCNSA-N
Synonym s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s
PubChem CID 6713753
IUPAC Name (2S)-butan-2-amine
SMILES CCC(C)N

Specifications

Density 0.722
pH 12.5
Boiling Point 63°C
Flash Point −19°C (−2°F)
Odor Amine-like
Refractive Index 1.393
Quantity 1 g
UN Number UN2733
Beilstein 1718760
Sensitivity Air Sensitive
Merck Index 14,1544
Solubility Information Fully miscible in water.
Optical Rotation +7.5° (Neat)
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Aminobutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'