missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminohexane, ChiPros 99+%, ee 99+%, Thermo Scientific™

£30.59 - £394.82

Chemical Identifiers

CAS 70492-67-0
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00671626
InChI Key WGBBUURBHXLGFM-UHFFFAOYNA-N
Synonym s-2-aminohexane, 2s-hexan-2-amine, s-2-hexylamine, s-+-2-aminohexane, s-+-2-aminohexane, chipros, ee 99+%, s-2-hexaneamine, pubchem6750, 2s-2-hexanamine, 2-hexanamine, 2s, +-s-2-aminohexane
PubChem CID 6999846
SMILES CCCCC(C)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344206
View Documents
Thermo Scientific Alfa Aesar
L19305.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11364206
View Documents
Thermo Scientific Alfa Aesar
L19305.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(+)-2-Aminohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water (17g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Specifications

Chemical Identifiers

70492-67-0
101.19
WGBBUURBHXLGFM-UHFFFAOYNA-N
6999846
C6H15N
MFCD00671626
s-2-aminohexane, 2s-hexan-2-amine, s-2-hexylamine, s-+-2-aminohexane, s-+-2-aminohexane, chipros, ee 99+%, s-2-hexaneamine, pubchem6750, 2s-2-hexanamine, 2-hexanamine, 2s, +-s-2-aminohexane
CCCCC(C)N

Specifications

70492-67-0
0.755
−3°C (26°F)
MFCD00671626
5237601
s-2-aminohexane, 2s-hexan-2-amine, s-2-hexylamine, s-+-2-aminohexane, s-+-2-aminohexane, chipros, ee 99+%, s-2-hexaneamine, pubchem6750, 2s-2-hexanamine, 2-hexanamine, 2s, +-s-2-aminohexane
WGBBUURBHXLGFM-UHFFFAOYNA-N
hexan-2-amine
6999846
≥99%
(S)-(+)-2-Aminohexane
<-20°C
116°C
C6H15N
UN2733
Air Sensitive
Soluble in water (17g/L at 20°C)
CCCCC(C)N
101.19
101.19
ee ≥99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 622-789-6

missing translation for 'tsca' : No

SDS
Documents

Documents

Product Certifications