missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Benzyl-1-Boc-piperazine, 97%, Thermo Scientific Chemicals

Product Code. 15459527
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169447-86-3
Molecular Formula C16H24N2O2
Molecular Weight (g/mol) 276.38
MDL Number MFCD03787923
InChI Key QKUHUJCLUFLGCI-AWEZNQCLSA-N
Synonym s-1-boc-2-benzylpiperazine, s-1-boc-2-benzyl-piperazine, s-tert-butyl 2-benzylpiperazine-1-carboxylate, tert-butyl 2s-2-benzylpiperazine-1-carboxylate, s-n1-boc-2-benzylpiperazine, 2 s-benzyl-piperazine-1-carboxylic acid tert-butyl ester, s-2-benzyl-piperazine-1-carboxylic acid tert-butyl ester, n1-boc-2-benzylpiperazine, s-1-n-boc-2-benzylpiperazine, s-2-benzyl-1-boc-piperazine
PubChem CID 17750441
IUPAC Name tert-butyl (2S)-2-benzylpiperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCNC[C@@H]1CC1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 276.38
Percent Purity 97%
Chemical Name or Material (S)-2-Benzyl-1-Boc-piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'