missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals
£16.48 - £211.00
Chemical Identifiers
CAS | 3350-13-8 |
---|---|
Molecular Formula | C29H47N3O6 |
Molecular Weight (g/mol) | 533.71 |
MDL Number | MFCD00237341 |
InChI Key | CYMIEBREXUYHGN-UHFFFAOYNA-N |
Synonym | dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha |
PubChem CID | 49853427 |
IUPAC Name | N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid |
SMILES | C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465565
|
Thermo Scientific Alfa Aesar
H62772.MD |
250 mg | |||||||
15445565
|
Thermo Scientific Alfa Aesar
H62772.03 |
1 g |
£54.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15455565
|
Thermo Scientific Alfa Aesar
H62772.06 |
5 g |
£211.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
3350-13-8 | |
533.71 | |
CYMIEBREXUYHGN-UHFFFAOYNA-N | |
49853427 | |
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
C29H47N3O6 | |
MFCD00237341 | |
dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha | |
N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid |
Specifications
3350-13-8 | |
C29H47N3O6 | |
dicyclohexylamine s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, z-dab boc-oh dcha, z-dab boc-oh?cha, z-dab boc-oh.dcha, n-a-z-n-?-boc-l-2,4-diaminobutyric aciddicyclohexylaminesalt, s-2-benzyloxycarbonylamino-4-boc-amino butyric acid dicyclohexylammonium salt, z-dab boc-oh.dcha na-z-n4-boc-2,4-diaminobutyric acid dicyclohexylamine salt, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2s-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha | |
CYMIEBREXUYHGN-UHFFFAOYNA-N | |
N-cyclohexylcyclohexanamine;(2S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid | |
49853427 | |
98% | |
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt |
White | |
MFCD00237341 | |
Slightly soluble in water. | |
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O | |
533.71 | |
553.7 | |
Powder |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only