missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-(Boc-amino)-4-phenylbutyric acid, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Descripción
(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Especificaciones
Melting Point | 76°C to 80°C |
Beilstein | 3653506 |
Solubility Information | Soluble in methanol. |
Formula Weight | 279.34 |
Percent Purity | 98% |
Quantity | 1 g |
Optical Rotation | +6.5° (c=2 in Ethanol) |
Chemical Name or Material | (S)-2-(Boc-amino)-4-phenylbutyric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'