missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Boc-amino)butyric acid, 95%, Thermo Scientific Chemicals

Product Code. 15400509
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34306-42-8
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037267
InChI Key PNFVIPIQXAIUAY-UHFFFAOYNA-N
Synonym boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid
PubChem CID 2755934
SMILES CCC(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 1 g
Beilstein 6801706
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Formula Weight 203.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (S)-2-(Boc-amino)butyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'