missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methoxypropanol, 97%, Thermo Scientific Chemicals

Product Code. 10726622
Click to view available options
Quantity:
1 mL
2.5 mL
10 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116422-39-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD01632593
InChI Key YTTFFPATQICAQN-BYPYZUCNSA-N
Synonym 2s-2-methoxypropan-1-ol, s-+-2-methoxypropanol, s-2-methoxypropan-1-ol, unii-6725gr28fg, 2-methoxy-1-propanol, 2s, 1-propanol, 2-methoxy-, 2s, 2s-2-methoxy-1-propanol, s-2-methoxy-1-propanol, 2-s-methoxypropan-1-ol, 1-propanol, 2-methoxy-, s
PubChem CID 642402
IUPAC Name (2S)-2-methoxypropan-1-ol
SMILES CC(CO)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9380g/mL
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OCH3)CH2OH
Refractive Index 1.4060 to 1.4080
Quantity 1 mL
Specific Gravity 0.938
Specific Rotation Condition + 39.20 (20.00°C c=1.66,CHCl3)
Specific Rotation + 39.20
Formula Weight 90.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Methoxypropanol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'