missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2-Phenylglycinol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.
Solubility
Soluble in chloroform, dimethylsulfoxide and methanol. Limited solubility in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with oxidizing agents. Store at -20°C.
(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.
Solubility
Soluble in chloroform, dimethylsulfoxide and methanol. Limited solubility in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with oxidizing agents. Store at -20°C.
Specifications
Specifications
Melting Point | 76°C to 78°C |
Quantity | 25 g |
Beilstein | 3196190 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in chloroform,dimethylsulfoxide and methanol. Limited solubility in water. |
Optical Rotation | +30° (c=0.75 in 1N HCl) |
Formula Weight | 137.18 |
Percent Purity | ≥98% |
Chemical Name or Material | (S)-(+)-2-Phenylglycinol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'