missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%, Thermo Scientific Chemicals
£81.90 - £222.00
Chemical Identifiers
CAS | 33878-99-8 |
---|---|
Molecular Formula | C11H14O2 |
Molecular Weight (g/mol) | 178.231 |
MDL Number | MFCD00213797 |
InChI Key | DNHDRUMZDHWHKG-NSHDSACASA-N |
Synonym | s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione # |
PubChem CID | 736069 |
IUPAC Name | (8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione |
SMILES | CC12CCC(=O)C=C1CCCC2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11388046
|
Thermo Scientific Alfa Aesar
L13334.ME |
500 mg |
£81.90
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398046
|
Thermo Scientific Alfa Aesar
L13334.04 |
2 g |
£222.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33878-99-8 | |
178.231 | |
DNHDRUMZDHWHKG-NSHDSACASA-N | |
736069 | |
CC12CCC(=O)C=C1CCCC2=O |
C11H14O2 | |
MFCD00213797 | |
s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione # | |
(8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione |
Specifications
33878-99-8 | |
C11H14O2 | |
1569979 | |
DNHDRUMZDHWHKG-NSHDSACASA-N | |
CC12CCC(=O)C=C1CCCC2=O | |
178.231 | |
178.23 | |
(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione |
48°C to 52°C | |
MFCD00213797 | |
s-+-3,4,8,8a-tetrahydro-8a-methyl-1,6 2h,7h-naphthalenedione, 8as-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-3,4,7,8-tetrahydro-2h-naphthalene-1,6-dione, s-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 8as-8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione, 8as-+-8a-methyl-3,4,8,8a-tetrahydronaphthalene-1,6 2h,7h-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl-, 8as, wieland-mieschler ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione # | |
+100° (c=1 in Benzene) | |
(8aS)-8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione | |
736069 | |
99% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only