missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine, 96%, Thermo Scientific™

Product Code. 15494006
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122536-72-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD06858462
InChI Key FPXJNSKAXZNWMQ-NSHDSACASA-N
Synonym s-1-cbz-3-aminopyrrolidine, s-+-1-cbz-3-aminopyrrolidine, benzyl 3s-3-aminopyrrolidine-1-carboxylate, s-3-amino-1-cbz-pyrrolidine, s-1-n-cbz-3-aminopyrrolidine, s-benzyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-pyrrolidine-1-carboxylic acid benzyl ester, s-1-benzyloxycarbonyl-3-aminopyrrolidine, s-3-amino-1-benzyloxycarbonyl pyrrolidine, s-+-3-amino-1-benzyloxycarbonyl pyrrolidine
PubChem CID 1519355
IUPAC Name benzyl (3S)-3-aminopyrrolidine-1-carboxylate
SMILES C1CN(CC1N)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 310°C to 316°C
Density 1.155
Flash Point >110°C (230°F)
Refractive Index 1.54
Quantity 1 g
Sensitivity Air Sensitive
Optical Rotation +5° (c=1 in Methanol)
Formula Weight 220.27
Percent Purity 96%
Chemical Name or Material (S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'