missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Beilstein | 7057792 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 426.47 |
Percent Purity | 95% |
Chemical Name or Material | (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'