missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-(Boc-amino)adipic acid 6-benzyl ester, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Starting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Starting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Starting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Beilstein | 8079168 |
Formula Weight | 351.4 |
Percent Purity | 95% |
Quantity | 1 g |
Chemical Name or Material | (S)-3-(Boc-amino)adipic acid 6-benzyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'