missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Fluoropyrrolidine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11451820
Click to view available options
Quantity:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 136725-53-6
Molecular Formula C4H9ClFN
Molecular Weight (g/mol) 125.571
MDL Number MFCD04038718
InChI Key LENYOXXELREKGZ-WCCKRBBISA-N
Synonym s-3-fluoropyrrolidine hydrochloride, s-+-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine hydrochloride, 3s-+-3-fluoropyrrolidine hydrochloride, s-3-fluoro-pyrrolidine hcl, s ;-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine, chloride, s-3-fluoropyrrolidine hcl
PubChem CID 16217739
IUPAC Name (3S)-3-fluoropyrrolidine;hydrochloride
SMILES C1CNCC1F.Cl

Spezifikation

Melting Point 183°C to 186°C
Quantity 5 g
Formula Weight 125.57
Percent Purity 97%
Chemical Name or Material (S)-(+)-3-Fluoropyrrolidine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'