missing translation for 'onlineSavingsMsg'
Learn More
(S)-3-Hydroxypyrrolidine hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 15479577

(S)-3-Hydroxypyrrolidine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15479577
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15479577

Brand: Thermo Scientific Alfa Aesar H52815.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122536-94-1
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD00272298
InChI Key QPMSJEFZULFYTB-WCCKRBBISA-N
Synonym s-3-hydroxypyrrolidine hydrochloride, s-pyrrolidin-3-ol hydrochloride, 3s-pyrrolidin-3-ol hydrochloride, s-+-3-pyrrolidinol hydrochloride, s-3-hydroxypyrrolidine-hcl, s-3-pyrrolidinol hydrochloride, 3s-pyrrolidinol hydrochloride, s-3-pyrrolidinol hcl, s-pyrrolidin-3-ol, hcl, s---3-pyrrolidinol hydrochlorideShow MoreShow Less
PubChem CID 22309122
IUPAC Name (3S)-pyrrolidin-3-ol;hydrochloride
SMILES C1CNCC1O.Cl

Specifications

Melting Point 106°C to 107°C
Boiling Point 108°C to 110°C (8 mmHg)
Quantity 5 g
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material (S)-3-Hydroxypyrrolidine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ26I046 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberH52815.06,H52815.03
Lot NumberP18H055 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52815.06,H52815.03
Lot NumberC21Z032 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52815.06,H52815.03
Lot NumberL12Y020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52815.06,H52815.03
Lot NumberG12X026 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52815.06,H52815.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-3-Hydroxypyrrolidine hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.