missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(1-Aminoethyl)phenol, 97%, Thermo Scientific Chemicals

Product Code. 15469878
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221670-72-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD03844646
InChI Key CDQPLIAKRDYOCB-UHFFFAOYNA-N
PubChem CID 41097924
IUPAC Name 4-[(1S)-1-aminoethyl]phenol
SMILES CC(N)C1=CC=C(O)C=C1

Specifications

Color White
Quantity 1 g
UN Number UN3259
Synonym 4-1s-1-aminoethyl phenol, s-4-1-aminoethyl phenol, phenol, 4-1s-1-aminoethyl, s-4-1-aminoethyl-phenol, s-4-1-amino-ethyl-phenol, s-4-1-amino-aethyl-phenol, phenol,4-1s-1-aminoethyl, s-4-hydroxy-alpha-methylbenzylamine, phenol, 4-1s-1-aminoethyl-9ci, s-4-hydroxy-? inverted exclamation marka-methylbenzylamine
Formula Weight 137.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (S)-4-(1-Aminoethyl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'