missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee), Thermo Scientific Chemicals

£88.50 - £281.00

Chemical Identifiers

CAS 54656-96-1
MDL Number MFCD00077866
InChI Key HVOAMIOKNARIMR-LURJTMIESA-N
Synonym s-1-pyridin-4-yl ethanol, s---1-4-pyridyl ethanol, 1s-1-pyridin-4-yl ethan-1-ol, 1s-1-pyridin-4-yl ethanol, s-1-4-pyridyl ethanol, 1s-1-4-pyridyl ethanol, s-1-4-pyridinyl ethanol, s--1-4-pyridyl ethanol, 1s-1-4-pyridinyl ethanol, --s-1-4-pyridyl ethanol
PubChem CID 10920507
IUPAC Name (1S)-1-pyridin-4-ylethanol
SMILES CC(C1=CC=NC=C1)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10404125
View Documents
Thermo Scientific Acros
393372500
250 mg Glass bottle
£88.50
250mg
Please to purchase this item. Need a web account? Register with us today!
10126753
View Documents
Thermo Scientific Acros
393370010
1 g Glass bottle
£281.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54656-96-1
HVOAMIOKNARIMR-LURJTMIESA-N
10920507
CC(C1=CC=NC=C1)O
MFCD00077866
s-1-pyridin-4-yl ethanol, s---1-4-pyridyl ethanol, 1s-1-pyridin-4-yl ethan-1-ol, 1s-1-pyridin-4-yl ethanol, s-1-4-pyridyl ethanol, 1s-1-4-pyridyl ethanol, s-1-4-pyridinyl ethanol, s--1-4-pyridyl ethanol, 1s-1-4-pyridinyl ethanol, --s-1-4-pyridyl ethanol
(1S)-1-pyridin-4-ylethanol

Specifications

54656-96-1
100.0
White
Authentic
C7H9NO
−58.50° (20°C c=1,chloroform)
s-1-pyridin-4-yl ethanol, s---1-4-pyridyl ethanol, 1s-1-pyridin-4-yl ethan-1-ol, 1s-1-pyridin-4-yl ethanol, s-1-4-pyridyl ethanol, 1s-1-4-pyridyl ethanol, s-1-4-pyridinyl ethanol, s--1-4-pyridyl ethanol, 1s-1-4-pyridinyl ethanol, --s-1-4-pyridyl ethanol
CC(C1=CC=NC=C1)O
123.155
123.15
Needles
99.0
67°C to 70°C
99.0% min.
99+%
MFCD00077866
-56° to -61° (20°C, 589nm) (C=1, chloroform)
HVOAMIOKNARIMR-LURJTMIESA-N
(1S)-1-pyridin-4-ylethanol
10920507
≥99.0%
(S)-4-(1-Hydroxyethyl)pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Identifier
  • (S)-4-(1-Hydroxyethyl)pyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only