missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-4-(4-Aminobenzyl)-2-oxazolidinone, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone is an intermediate of zolmitriptan (sc-220415) which acts as a novel inhibitor of the cytochrome P-450 enzyme aromatase.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Keep container tightly closed.
(S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone is an intermediate of zolmitriptan (sc-220415) which acts as a novel inhibitor of the cytochrome P-450 enzyme aromatase.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store at -20°C. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
Specifications
Melting Point | 107°C to 111°C |
Quantity | 25 g |
Solubility Information | Soluble in dimethyl sulfoxide and methanol. |
Formula Weight | 192.22 |
Percent Purity | 97% |
Chemical Name or Material | (S)-4-(4-Aminobenzyl)-2-oxazolidinone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'