missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Benzyl-2-oxazolidinone, 99%, Thermo Scientific Chemicals

Product Code. 11433767
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(S)-(-)-4-Benzyl-2-oxazolidinone is used in chiral auxiliary asymmetric alkylation. It has been used in the synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides. It was used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-4-Benzyl-2-oxazolidinone is used in chiral auxiliary asymmetric alkylation. It has been used in the synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides. It was used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90719-32-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00064496
InChI Key OJOFMLDBXPDXLQ-VIFPVBQESA-N
Synonym s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
PubChem CID 736225
IUPAC Name (4S)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@@H](CC2=CC=CC=C2)CO1

Specifications

Melting Point 87°C to 89°C
Beilstein 3649667
Solubility Information Insoluble in water.
Quantity 1 g
Formula Weight 177.2
Optical Rotation −64° (c=1 in Chloroform)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material (S)-(-)-4-Benzyl-2-oxazolidinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.