missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Carboxyphenylglycine, Tocris Bioscience™

Codice prodotto. 30001077
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Questo articolo non è restituibile. Consulta la politica di reso
Questo articolo non è restituibile. Consulta la politica di reso

Competitive group I mGlu antagonist; also weak group II agonist

(S)-4-Carboxyphenylglycine is a competitive group I metabotropic glutamate receptor antagonist, with selectivity for mGlu1a/1a over mGlu5a/5b.

Identificatori chimici

CAS 134052-73-6
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
InChI Key VTMJKPGFERYGJF-ZETCQYMHSA-N
Synonym s-4-carboxyphenylglycine, s-4cpg, chembl94990, s-4-amino carboxy methyl benzoic acid, 4-1s-1-amino-2-hydroxy-2-oxo-ethyl benzoic acid, 4-1s-1-amino-2-hydroxy-2-oxoethyl benzoic acid, 4-amino-carboxy-methyl-benzoic acid, tocris-0323, 4-carboxy-l-phenylglycine
PubChem CID 5311459
IUPAC Name 4-[(S)-amino(carboxy)methyl]benzoic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)C(=O)O

Specifica

Formula Weight 195.17
Percent Purity >99%
Quantity 10 mg
Chemical Name or Material (S)-4-Carboxyphenylglycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'