missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-4-Carboxyphenylglycine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
(S)-4-Carboxyphenylglycine is a competitive group I metabotropic glutamate receptor antagonist, with selectivity for mGlu1a/1a over mGlu5a/5b.
Specifications
Specifications
Formula Weight | 195.17 |
Percent Purity | >99% |
Quantity | 50 mg |
Chemical Name or Material | (S)-4-Carboxyphenylglycine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'