missing translation for 'onlineSavingsMsg'
Learn More
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%, Thermo Scientific Chemicals Product Code.: 11456850

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%, Thermo Scientific Chemicals

Product Code. 11456850
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11456850

Brand: Thermo Scientific Alfa Aesar L18359.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17342-08-4
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00077792
InChI Key HOBJEFOCIRXQKH-BYPYZUCNSA-N
Synonym l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847Show MoreShow Less
PubChem CID 643511
IUPAC Name (5S)-5-(hydroxymethyl)pyrrolidin-2-one
SMILES C1CC(=O)NC1CO

Specifications

Melting Point 78°C to 82°C
Boiling Point 147°C to 149°C (0.06 mmHg)
Quantity 5 g
Beilstein 4657914
Solubility Information Soluble in water.
Optical Rotation +30° (c=2 in Ethanol)
Formula Weight 115.13
Percent Purity 98%
Chemical Name or Material (S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10171673 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog NumberL18359.06,L18359.03
Lot NumberUSLY001124 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL18359.06,L18359.03
Lot NumberH17R037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL18359.06,L18359.03
Lot Number10171673 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL18359.06,L18359.03
Lot NumberL13W030 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL18359.06,L18359.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.