missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Safety Waste Cap, S70/71, Type 3
GREENER_CHOICE

Product Code. 10580241 Shop All SCAT Europe Products
Click to view available options
Stopper Number:
S70/71
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10580241

Brand: SCAT™ 107926

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Safety Waste Caps for the safe disposal of liquid waste. For maximum protection and efficient work with hazardous waste liquids. Offers multiple options to connect different tubing and capillary sizes.

  • 360° free turning PTFE body, resistant against aggressive media
  • Exhaust filter system against harmful vapors
  • Fittings and tube connectors in different sizes
  • Flexible system
  • Time saving in changing the waste containers while tubes are fixed
  • Lab safety, technical protective measures, optimal protection when collecting liquid waste
  • Cost efficiency
  • Huge range of accessories, optional: funnel use, level control preventing overflowing containers
  • Huge portfolio of thread sizes suitable for waste containers from different manufacturers
  • Safety disposal of HPLC waste
  • High work performance
  • Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
  • Sealed bottles and containers
  • Create a comfortable working climate

Specifications

For Use With (Equipment) Waste Disposal Container
Material PTFE
Number of Channels 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID
For Use With (Application) HPLC Disposal Systems
Type Safety Waste Cap
Color Black
Stopper Number S70/71
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.