Learn More
SCAT™ Safety Waste Caps
Safety Waste Caps for the safe disposal of liquid waste. For maximum protection and efficient work with hazardous waste liquids. Offers multiple options to connect different tubing and capillary sizes.
Brand: SCAT™ 108023
97.54 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.45000kg
Description
- 360° free turning PTFE body, resistant against aggressive media
- Exhaust filter system against harmful vapors
- Fittings and tube connectors in different sizes
- Flexible system
- Time saving in changing the waste containers while tubes are fixed
- Lab safety, technical protective measures, optimal protection when collecting liquid waste
- Cost efficiency
- Huge range of accessories, optional: funnel use, level control preventing overflowing containers
- Huge portfolio of thread sizes suitable for waste containers from different manufacturers
- Safety disposal of HPLC waste
- High work performance
- Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
- Sealed bottles and containers
- Create a comfortable working climate
Specifications
3 x 2.3/3.2 mm OD, Connector for Exhaust Filter | |
S50 | |
Black | |
Waste Disposal Container |
HPLC Disposal Systems | |
Safety Waste Cap | |
PTFE |