missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ Safety Waste Caps
Safety Waste Caps for the safe disposal of liquid waste. For maximum protection and efficient work with hazardous waste liquids. Offers multiple options to connect different tubing and capillary sizes.
Brand: SCAT™ 107947
124.39 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 360° free turning PTFE body, resistant against aggressive media
- Exhaust filter system against harmful vapors
- Fittings and tube connectors in different sizes
- Flexible system
- Time saving in changing the waste containers while tubes are fixed
- Lab safety, technical protective measures, optimal protection when collecting liquid waste
- Cost efficiency
- Huge range of accessories, optional: funnel use, level control preventing overflowing containers
- Huge portfolio of thread sizes suitable for waste containers from different manufacturers
- Safety disposal of HPLC waste
- High work performance
- Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
- Sealed bottles and containers
- Create a comfortable working climate
Specifications
Not Autoclavable | |
HPLC Disposal Systems | |
4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter | |
S90 |
Black | |
Waste Disposal Container | |
PTFE, Polypropylene, PFA, HDPE | |
Safety Waste Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCAT™ Safety Waste Caps > Safety Waste Caps; Size: S90; Includes: 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter
Spot an opportunity for improvement?Share a Content Correction