missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph Instruments S-Box Package 1
Shop All Heidolph Instruments Products
Click to view available options
Color:
Blue
Green
Orange
Violet
Unit Size:
Each
Description
- Each S-box is a stand-alone miniature incubator.
- Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
- The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
- Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
- To avoid mix-up, the front cover of the S-box is available in different colors.
Specifications
Specifications
| Color | Orange |
| Compressor | Orange |
| Concentration Range | Blue |
| Content And Storage | Violet |
| Includes | 1 S-box, 1 Tool Kit, 4 inner boxes, 4 water bags, 4 front covers |
| Input Water Pressure | Orange |
| Installation | Blue |
| Insulation | Violet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction