Learn More
Heidolph Instruments S-Box Package 2
The S-Box Package 2 includes additional accessories to complete an existing S-Biosystem incubation system.
Brand:  Heidolph Instruments 400-03003-00
 View more versions of this product
Product Code. 17838000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
Specifications
Specifications
Violet
1 S-box, 1 tool kit, 8 inner boxes, 8 water bags, 4 front covers