Learn More
Asecos™ S-CLASSIC-90 1-Door Light Gray Safety Storage Cabinets
Safety storage cabinets for unrestricted storage of flammable hazardous materials in work areas in accordance with EN 14470-1 and TRGS 510.
Brand: Asecos™ 3023800133735
1916.82 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 230.00000kg
Description
- Aseco’s first EN-compliant safety storage cabinet
- Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
- Available in a range of widths and heights
- Versatile fields of application due to various interior equipment
- Convenient handling – door operation with minimal effort
- Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
- Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- In accordance with EN 14470-1 and TRGS 510
Specifications
2 x 75 kg sheet steel shelves; 1 x 75 kg sheet steel insert; 1 x 22 L sheet steel sump | |
Storage of flammable hazardous materials | |
Steel | |
616 x 596 x 1298 mm | |
Light Gray | |
Triple-hinged | |
EN 14470-1, TRGS 510 |
Safety storage cabinet | |
Powder-coated | |
Without door open arrest system | |
202.5 kg | |
Shelves 75 kg each; Insert 75 kg; Sump 22 L | |
1 |