missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-CLASSIC-90 1-Door Light Gray Safety Storage Cabinets with Door Open Arrest System
Safety storage cabinets for unrestricted storage of flammable hazardous materials in work areas in accordance with EN 14470-1 and TRGS 510.
£2388.18
Specifications
Certifications/Compliance | EN 14470-1, TRGS 510 |
---|---|
Color | Light Gray |
Dimensions (D x W x H) | 616 x 596 x 1968 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
15919371
|
Asecos™
3022800133659 |
3 x 75 kg sheet steel shelves; 1 x 75 kg sheet steel insert; 1 x 22 L sheet steel sump |
£2388.18
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Aseco’s first EN-compliant safety storage cabinet
- Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
- Available in a range of widths and heights
- Versatile fields of application due to various interior equipment
- Convenient handling – door operation with minimal effort
- Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
- Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- In accordance with EN 14470-1 and TRGS 510
Specifications
EN 14470-1, TRGS 510 | |
616 x 596 x 1968 mm | |
Safety storage cabinet | |
Storage of flammable hazardous materials | |
1 |
Light Gray | |
Steel | |
Triple-hinged | |
Left-hinged door open arrest system | |
Powder-coated |