missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ S-CLASSIC-90 2-Door Light Gray Safety Storage Cabinets without Door Arrest System

For the safe and approved storage of flammable hazardous substances in working areas, with an optional door open arrest system.

Brand:  Asecos™ 3011500133715

Additional Details : Weight : 453.00000kg

Product Code. 15526855

  • £2921.89 / Each
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Aseco’s first EN-compliant safety storage cabinet
  • Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
  • Available in a range of widths and heights
  • Versatile fields of application due to various interior equipment
  • Convenient handling – door operation with minimal effort
  • Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
  • Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
  • Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • Fire resistance of 90 minutes
  • Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • In accordance with EN 14470-1 and TRGS 510
Specifications

Specifications

2 x sheet steel shelves; 1 x sheet steel perforated insert; 1 x sheet steel sump
Storage of flammable and hazardous substances
Steel
616 x 1196 x 1968 mm
Light Gray
Triple-hinged
EN 14470-1
Safety storage cabinet
Powder-coated
Without door open arrest system
451.5 kg
Shelves 75 kg each; Insert 75 kg; Sump 33 L
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers