missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-CLASSIC-90 One-Door Yellow Safety Storage Cabinets
Safety storage cabinets for unrestricted storage of flammable hazardous materials in work areas in accordance with EN 14470-1 and TRGS 510.
£2160.00
Specifications
Certifications/Compliance | EN 14470-1, TRGS 510 |
---|---|
Color | Yellow |
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Triple-hinged |
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
15891518
|
Asecos™
3023800233735 |
2 x Sheet Steel Shelves: 75 kg, Sheet Steel Insert: 75 kg, Sheet Steel Sump: 22 L, Max load: 600 kg |
£2160.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Aseco’s first EN-compliant safety storage cabinet
- Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
- Available in a range of widths and heights
- Versatile fields of application due to various interior equipment
- Convenient handling – door operation with minimal effort
- Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
- Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- In accordance with EN 14470-1 and TRGS 510
Specifications
EN 14470-1, TRGS 510 | |
Steel | |
Triple-hinged | |
1 |
Yellow | |
Safety storage cabinet | |
Storage of flammable hazardous materials | |
Powder-coated |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Asecos™ S-CLASSIC-90 One-Door Yellow Safety Storage Cabinets