missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ S-CLASSIC-90 One-Door Yellow Safety Storage Cabinets with Door Open Arrest System

Safety storage cabinets for unrestricted storage of flammable hazardous materials in work areas in accordance with EN 14470-1 and TRGS 510.

Brand:  Asecos™ 3022800233681

Additional Details : Weight : 260.00000kg

 View more versions of this product

Product Code. 15778362

  • £2739.04 / Each
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Aseco’s first EN-compliant safety storage cabinet
  • Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
  • Available in a range of widths and heights
  • Versatile fields of application due to various interior equipment
  • Convenient handling – door operation with minimal effort
  • Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
  • Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
  • Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • Fire resistance of 90 minutes
  • Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • In accordance with EN 14470-1 and TRGS 510
Specifications

Specifications

4 x 25 kg/5 L sheet steel drawers
Storage of flammable hazardous materials
Steel
616 x 596 x 1968 mm
Yellow
Triple-hinged
EN 14470-1, TRGS 510
Safety storage cabinet
Powder-coated
Left-hinged door open arrest system
291 kg
4 x Drawers: 25 kg/5 L each, Max load: 600 kg
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers