missing translation for 'onlineSavingsMsg'
Learn More
S-Ethyl thiopropionate, 97%, Thermo Scientific™ Product Code.: 11471407

S-Ethyl thiopropionate, 97%, Thermo Scientific™

Product Code. 11471407
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11471407

Brand: Thermo Scientific Alfa Aesar B24918.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-42-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00027016
InChI Key HNEVHBHRLCAKKQ-UHFFFAOYSA-N
Synonym s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester
PubChem CID 75513
IUPAC Name S-ethyl propanethioate
SMILES CCC(=O)SCC

Specifications

Density 0.958
Boiling Point 137°C to 138°C
Flash Point 27°C (80°F)
Odor Stench
Refractive Index 1.459
Quantity 100 g
UN Number UN1993
Beilstein 1740740
Formula Weight 118.2
Percent Purity 97%
Chemical Name or Material S-Ethyl thiopropionate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S-Ethyl thiopropionate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.