missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Glycidyl trityl ether, 98%, ee 98%, Thermo Scientific™

Product Code. 15465747
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15465747

Brand: Thermo Scientific Alfa Aesar H32961.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129940-50-7
Molecular Formula C22H20O2
Molecular Weight (g/mol) 316.4
MDL Number MFCD00723373
InChI Key XFSXUCMYFWZRAF-NRFANRHFSA-N
Synonym s---trityl glycidyl ether, s-2-trityloxy methyl oxirane, s-trityl glycidyl ether, s---glycidyl trityl ether, s-glycidyl trityl ether, 2s-2-trityloxymethyl oxirane, s-tritylglycidylether, s-2-triphenylmethoxymethyl oxirane, s---glycidyl triphenylmethyl ether, s-glycidyl triphenylmethyl ether
PubChem CID 7168113
IUPAC Name (2S)-2-(trityloxymethyl)oxirane
SMILES C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 99°C to 102°C
Quantity 5 g
Optical Rotation −10.5° (c=1 in Chloroform)
Formula Weight 316.39
Percent Purity 98%
Assay ee 98%
Chemical Name or Material (S)-(-)-Glycidyl trityl ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.