missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Ibuprofen, 99%, Thermo Scientific Chemicals

Product Code. 10193091 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10193091

Brand: Thermo Scientific Acros 333200010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(S)-(+)-Ibuprofen, CAS # 51146-56-6, is the active enantomerof the non-steroidal anti-inflammatory drug, ibuprofen.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ibuprofen is a nonsteroidal anti-inflammatory drug commonly used in its racemic form
  • (S)-(+)-Ibuprofen is the active enantiomer of this drug that confers anti-inflammatory activity
  • (S)-(+)-Ibuprofen inhibits prostaglandin synthesis and thus reduces inflammation and pain when ingested

Applications

  • As the active enantiomer of ibuprofen, (S)-(+)-Ibuprofen is used in laboratory research for pharmacokinetic studies
  • (S)-(+)-Ibuprofen may be used as an inhibitor of cycloogenase 1 and 2 and an agonist of PPARalpha and PPARgamma
  • (S)-(+)-Ibuprofen may be used as material to study development of drug delivery systems
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51146-56-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.28
MDL Number MFCD00069289
InChI Key HEFNNWSXXWATRW-JTQLQIEISA-N
Synonym s-+-ibuprofen, dexibuprofen, s +-ibuprofen, s-2-4-isobutylphenyl propanoic acid, s-ibuprofen, d-ibuproten, s-+-2-4-isobutylphenyl propionic acid, s-+-4-isobutyl-alpha-methylphenylacetic acid, +-s-p-isobutylhydratropic acid, seractil
PubChem CID 39912
ChEBI CHEBI:43415
IUPAC Name (2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 49.0°C to 53.0°C
Color White
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition + 57.00 (20.00°C c=2,EtOH)
Specific Rotation + 57.00
Solubility Information Solubility in water: insoluble. Other solubilities: readily soluble in most organic solvents
Formula Weight 206.28
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(+)-Ibuprofen, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-Ibuprofen
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : (S)-(+)-Ibuprofen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.