missing translation for 'onlineSavingsMsg'
Learn More
(S)-(+)-Ibuprofen, 99%, Thermo Scientific Chemicals Product Code.: 10285271

(S)-(+)-Ibuprofen, 99%, Thermo Scientific Chemicals

Product Code. 10285271
5 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10285271

Brand: Thermo Scientific Acros 333200050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(S)-(+)-Ibuprofen, CAS # 51146-56-6, is the active enantomerof the non-steroidal anti-inflammatory drug, ibuprofen.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ibuprofen is a nonsteroidal anti-inflammatory drug commonly used in its racemic form
  • (S)-(+)-Ibuprofen is the active enantiomer of this drug that confers anti-inflammatory activity
  • (S)-(+)-Ibuprofen inhibits prostaglandin synthesis and thus reduces inflammation and pain when ingested

Applications

  • As the active enantiomer of ibuprofen, (S)-(+)-Ibuprofen is used in laboratory research for pharmacokinetic studies
  • (S)-(+)-Ibuprofen may be used as an inhibitor of cycloogenase 1 and 2 and an agonist of PPARalpha and PPARgamma
  • (S)-(+)-Ibuprofen may be used as material to study development of drug delivery systems
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51146-56-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.28
MDL Number MFCD00069289
InChI Key HEFNNWSXXWATRW-JTQLQIEISA-N
Synonym s-+-ibuprofen, dexibuprofen, s +-ibuprofen, s-2-4-isobutylphenyl propanoic acid, s-ibuprofen, d-ibuproten, s-+-2-4-isobutylphenyl propionic acid, s-+-4-isobutyl-alpha-methylphenylacetic acid, +-s-p-isobutylhydratropic acid, seractilShow MoreShow Less
PubChem CID 39912
ChEBI CHEBI:43415
IUPAC Name (2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 49.0°C to 53.0°C
Color White
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition + 57.00 (20.00°C c=2,EtOH)
Specific Rotation + 57.00
Solubility Information Solubility in water: insoluble. Other solubilities: readily soluble in most organic solvents
Formula Weight 206.28
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(+)-Ibuprofen, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460712 Certificate TypeCertificate of Analysis Date29/02/2024 Catalog Number333200050,333200010
Lot NumberA0448846 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number333200050,333200010
Lot NumberA0406513 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number333200050,333200010
Lot NumberA0415278 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number333200050,333200010
Lot NumberA0437047 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number333200050,333200010
compliance-icons
Product Identifier
  • (S)-(+)-Ibuprofen
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : (S)-(+)-Ibuprofen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(+)-Ibuprofen, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.