missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-Mandelic Acid ≥99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 130°C to 135°C |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Color | Beige-Brown to White |
Assay Percent Range | 99+% |
Linear Formula | C6H5CH(OH)CO2H |
Beilstein | 10, 192 |
Formula Weight | 152.15 |
Percent Purity | ≥99.0% (Acid) |
Quantity | 25 g |
Show More |
Safety and Handling
Product Identifier
- Lead(II) acetate trihydrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'