missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Mandelic acid, 99+%

Product Code. 10396810 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10396810

Brand: Thermo Scientific Acros 125301000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17199-29-0 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17199-29-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004495
InChI Key IWYDHOAUDWTVEP-ZETCQYMHSA-N
Synonym s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
PubChem CID 439616
ChEBI CHEBI:32800
IUPAC Name (2S)-2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

Melting Point 130°C to 135°C
CAS Min % 99.0
CAS Max % 100.0
Color Beige-Brown to White
Assay Percent Range 99+%
Linear Formula C6H5CH(OH)CO2H
Beilstein 10, 192
Formula Weight 152.15
Percent Purity ≥99.0% (Acid)
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-Mandelic acid, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lead(II) acetate trihydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.