missing translation for 'onlineSavingsMsg'
Learn More

(S)-MCPG, Tocris Bioscience™

Product Code. 30001081
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective mGlu antagonist; active isomer of (RS)-MCPG (Cat. No. 0336)

Active isomer of (RS)-MCPG . Non-selective group I/group II metabotropic glutamate receptor antagonist.

Available as part of the Group I mGlu Receptor Tocriset™, Group II mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . Racemate also available.

Chemical Identifiers

CAS 150145-89-4
Molecular Formula C10H10NO4
Molecular Weight (g/mol) 208.19
MDL Number MFCD00216858
InChI Key DNCAZYRLRMTVSF-JTQLQIEISA-M
Synonym 4-1s-1-ammonio-1-carboxylatoethyl benzoate
PubChem CID 7019268
IUPAC Name 4-[(1S)-1-azaniumyl-1-carboxylatoethyl]benzoate
SMILES C[C@@]([NH3+])(C([O-])=O)C1=CC=C(C=C1)C([O-])=O

Specifications

Formula Weight 209.2
Percent Purity >99%
Quantity 10 mg
Chemical Name or Material (S)-MCPG
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'