missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-α-Ethylalanine, 98%, 98% ee, Thermo Scientific Chemicals

Product Code. 11955521
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151171-11-8
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD12031697
InChI Key SHZXLTCEPXVCSV-JTQLQIEISA-N
Synonym boc-l-isovaline, s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, s-2-tert-butoxycarbonylamino-2-methylbutanoic acid, n-1,1-dimethylethyl oxy carbonyl-d-isovaline, s-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
PubChem CID 14284792
IUPAC Name (2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C)(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 104°C to 112°C
CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation 12
Formula Weight 217.27
Percent Purity 98%,98% ee
Quantity 100 mg
Physical Form Powder
Chemical Name or Material (S)-N-BOC-α-Ethylalanine, 98% ee
Show More Show Less
Product Identifier
  • (S)-N-BOC-alpha-Ethylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'