missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-N-BOC-Propargylglycine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 85.5°C |
Color | White to Yellow |
Enantiomeric Excess | 97.5% min. (GC) |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Quantity | 500 mg |
Solubility Information | Solubility in water: soluble. |
Show More |
Safety and Handling
Product Identifier
- (S)-N-BOC-Propargylglycine
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'