missing translation for 'onlineSavingsMsg'
Learn More
S-n-Propyl thioacetate, 98+%, Thermo Scientific™ Product Code.: 11474937

S-n-Propyl thioacetate, 98+%, Thermo Scientific™

Product Code. 11474937
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11474937

Brand: Thermo Scientific Alfa Aesar B21875.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2307-10-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.19
MDL Number MFCD00039937
InChI Key SBWFWBJCYMBZEY-UHFFFAOYSA-N
Synonym s-propyl thioacetate, propyl thioacetate, ethanethioic acid, s-propyl ester, propyl thiolacetate, s-n-propyl thioacetate, acetic acid, thio-, s-propyl ester, unii-y7284nz6ep, 1-propylsulfanyl ethanone, n-propyl thio acetate, fema no. 3385
PubChem CID 61295
IUPAC Name S-propyl ethanethioate
SMILES CCCSC(C)=O

Specifications

Density 0.971
Boiling Point 137°C to 139°C
Refractive Index 1.46
Quantity 25 g
UN Number UN3272
Beilstein 1740765
Formula Weight 118.2
Percent Purity ≥98%
Chemical Name or Material S-n-Propyl thioacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10198541 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10173988 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberG7772A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberG7773A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10011876 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S-n-Propyl thioacetate, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.