Learn More
Fisherbrand™ S-Series Heated Ultrasonic Cleaning Bath
With state of the art microprocessor controlled ultrasonic cleaning and sweep technology. Fisherbrand™ S-Series Ultrasonic Cleaners offer a comprehensive range of analogue ultrasonic water baths covering the requirements for laboratory research, sonochemistry and degassing applications.
£301.82 - £4485.00
Specifications
Temperature (Metric) Range 30°C to 80°C
View More Specs
Products 13
Product Code Brand Capacity (Metric) Dimensions (D x W x H) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Dimensions (D x W x H) Price Quantity & Availability  
10591203
View accessory products View Documents Promotion Details
Fisherbrand™
1001214
0.8 L 116 x 206 x 178 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10551783
View accessory products View Documents Promotion Details
Fisherbrand™
1002060
1.75 L 180 x 175 x 212 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10162372
View accessory products View Documents Promotion Details
Fisherbrand™
1001955
2.75 L 179 x 300 x 214 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10215332
View accessory products View Documents Promotion Details
Fisherbrand™
1002006
4.25 L 179 x 300 x 264 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10345822
View accessory products View Documents Promotion Details
Fisherbrand™
1002238
5.75 L 186 x 365 x 264 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10395912
View accessory products View Documents Promotion Details
Fisherbrand™
1002140
6.9 L 179 x 568 x 214 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10663113
View accessory products View Documents Promotion Details
Fisherbrand™
1002170
9.4 L 179 x 568 x 264 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10239491
View accessory products View Documents Promotion Details
Fisherbrand™
1002279
9.5 L 278 x 365 x 264 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10204762
View accessory products View Documents Promotion Details
Fisherbrand™
1002825
12.75 L 278 x 365 x 321 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10102762
View accessory products View Documents Promotion Details
Fisherbrand™
1002877
18 L 340 x 390 x 321 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15280147
View accessory products View Documents Promotion Details
Fisherbrand™
1071162
28 L 340 x 568 x 321 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10366012
View accessory products View Documents Promotion Details
Fisherbrand™
1003908
45 L 370 x 615 x 467 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10427912
View accessory products View Documents Promotion Details
Fisherbrand™
1003946
90 L 570 x 715 x 467 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Long lifespan from cavitation resistant stainless steel tanks
  • LED display showing set and remaining time and pre-set and actual temperature
  • Newly developed cover reduces noise and accelerates the heating up process
  • Easy draining of cleaning liquid though the drain duct via a dial on the side
  • Operating panel arranged so that liquid cannot enter the electronics
  • Degas function key
Specifications
Specifications
30°C to 80°C