Learn More
Fisherbrand™ S-Series Unheated Ultrasonic Cleaning Bath
Designed with state of the art microprocessor controlled ultrasonic cleaning and sweep technology. Fisherbrand™ S-Series Ultrasonic Cleaners offer a comprehensive range of analogue ultrasonic baths covering the requirements for laboratory research, sonochemistry and degassing applications.
Brand:  Fisherbrand™
Code : 46
Additional Details : Weight : 13.80000kg
 View more versions of this product
Product Code. 10485982
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Long lifespan from cavitation resistant stainless steel tanks
  • LED display showing set and remaining time and pre-set and actual temperature
  • Ergonomically shaped plastic carrying handles
  • Newly developed cover reduces noise and accelerates the heating up process
  • Easy draining of cleaning liquid though the drain duct via a dial on the side
  • Operating panel arranged so that liquid cannot enter the electronics
  • Degas function key
Specifications
Specifications
V2A Stainless Steel
28 L
220 to 240VAC, 1500w
Elmasonic S 300 (H)
11 kg
220/240 VAC
340 x 568 x 321 mm
Laboratory, medical sector, dental laboratory, jewellery, watch sector, optician, workshop and service, industry
30°C to 80°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products