missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts S Test
Microgen Bioproducts S - Test - Based on the original method of Itano 1953. Intended for use as a screening reagent for the detection of Hb-S.
Marke: Novacyt - Microgen Bioproducts M96-50
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- For the in vitro screening of human blood samples to detect the presence of Haemoglobin-S, which is associated with the Sickle Cell trait.
Spezifikation
ISO13485 ISO9001 | |
Yes | |
Hb-S | |
Plasma | |
Screening Test | |
Positive and Negative Control Samples, Pipettes, Clear Plastic or Glasstest Tubes (75 x 12 mm), Timer, Centrifuge Capable of Spinning Tubes (at 1200 g (for Spin Technique Only), Latex Gloves, Safety Glasses and Other Appropriate Protective Garments, Biohazardous Waste Containers) | |
Polyethylene Dropper Bottles | |
Animal | |
5 Bottles (20 mL) and 5 vials (2.2 g) | |
Haemoglobin S | |
Eukaryota |
2°C to 8°C | |
Non-waived | |
5 Bottles (20 mL) and 5 vials (2.2 g) | |
At low levels of Hb-S (10-20%), the turbidity result may be equivocal and require confirmation by the centrifugation technique. Below 10% Hb-S, results of all solubility tests including S-Test are likely to be equivocal or negative.However, this level of sensitivity is sufficient for the diagnosis of cases of sickle cell trait with a low percentage of haemoglobin-S due to coexisting thalassaemia. | |
In Vitro Screening of Human Blood Samples to Detect The Presence of HaemoglobinS Which is Associated with Sickle Cell Trait | |
50 tests | |
3 per test | |
Carton | |
M96A 5 x 20 mL - Phosphate Buffer Solution, M96B 5 x 2.2 g - dry powder | |
Reliable results can be obtained with S-Test down to approximately 10% Hb-S. | |
3-5 minutes |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur