missing translation for 'onlineSavingsMsg'
Learn More
s-Trioxane, 99+%, Thermo Scientific Chemicals Product Code.: 10636152

s-Trioxane, 99+%, Thermo Scientific Chemicals

Product Code. 10636152
500 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10636152

Brand: Thermo Scientific Acros 140295000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-88-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00006563
InChI Key BGJSXRVXTHVRSN-UHFFFAOYSA-N
Synonym s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane
PubChem CID 8081
ChEBI CHEBI:38043
IUPAC Name 1,3,5-trioxane
SMILES C1OCOCO1

Specifications

Melting Point 61.0°C to 62.0°C
Color Colorless to White
Density 1.1700g/mL
Boiling Point 114.0°C to 116.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 19,381
Merck Index 15,9906
Specific Gravity 1.17
Solubility Information Solubility in water: soluble in water: 221g/L (25°C). Other solubilities: soluble in alcohols,ether,ketones,acetone,,chlorinatid and aromatic hydrocarbons,and,other organic solvents,slightly soluble in pentane and petr.ether
Sublimation Point 0115.0°C
Formula Weight 90.08
Percent Purity 99+%
Physical Form Crystalline Flakes or Crystals
Chemical Name or Material s-Trioxane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • s-Trioxane
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H335-May cause respiratory irritation.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : 1,3,5-Trioxane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
s-Trioxane, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.