missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-verbenone, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
Density | 0.9700g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 5 g |
Specific Rotation Condition | − 220.00 (20.00°C c=3−4,CHCL3) |
Formula Weight | 150.22 |
Percent Purity | 97% |
Chemical Name or Material | (S)-Verbenone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'