missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-WAY 100135 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective 5-HT1A receptor antagonist (IC50 = 15 nM). Selective over 5-HT1B, 1C, 2,α1, α2 and D2 receptors (IC50 > 1000 nM). Centrally active on systemic administration.
Specifications
Quantity | 10 mg |
Formula Weight | 468.47 |
Percent Purity | >99% |
Chemical Name or Material | (S)-WAY 100135 dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'