missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A12 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547056
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
S100A12 Polyclonal antibody specifically detects S100A12 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
S100A12 | |
Polyclonal | |
Unconjugated | |
S100A12 | |
CAAF1; CAGC; Calcitermin; calcium-binding protein in amniotic fluid 1; calgranulin C; calgranulin-C; CGRP; EN RAGE; ENRAGE; EN-RAGE; Extracellular newly identified RAGE-binding protein; migration inhibitory factor-related protein 6; MRP6; MRP-6; Neutrophil S100 protein; p6; Protein S100 A12; protein S100-A12; S100 calcium binding protein A12; S100 calcium-binding protein A12; S100 calcium-binding protein A12 (calgranulin C); S100A12 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6283 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P80511 | |
S100A12 | |
E. coli-derived recombinant human EN-RAGE/S100A12 Met1-Glu92 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |