Learn More
Thermo Scientific™ S110-AT Fixed Angle Rotor
Reduce separation time, processing samples 17% faster than previous similar rotors at an RCF of 691,000 x g using this fixed angle rotor.
Brand:  Thermo Scientific™ 45539
Product Code. 12111660
  • £20035.00 / Each
Explore more special offers
Description
Description
Reduce separation time, processing samples 17% faster than previous similar rotors at an RCF of 691,000 x g using the Thermo Scientific™ S110-AT Fixed Angle Rotor.
Compatible with:
Thermo Scientific™ Sorvall™ MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S110-AT Fixed Angle Rotor
Fixed Angle Rotor
690,652 x g
51.1 mm
15.4
13 mm
Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges
510,893 x g
Titanium
8 x 5 mL
110,000 rpm
24.5 mm
63 mm
2.4 lb. (1.07 kg)
30°
331,135 x g