missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ S3100 Sign and Label Printer- F&B Kit AZ
Increase the safety and efficiency of your food or beverage plant with this complete safety sign and label printer kit.
Brand: Brady™ 311117
Additional Details : Weight : 8.80760kg
Description
- Easily design and print professional food or beverage safety signs, pipe markers, and labels when you need them
- Print incredibly fast with automatic setup, built-in keyboard, and touchscreen interface
- Create reliable labels that are washdown resistant and do not crumble when exposed to cleaning agents or pressure
- Fully protect food processing with metal detectable signs that enable additional protection against contamination
Specifications
LCD Touchscreen | |
12 in. | |
9.5 in. | |
10 in. | |
Not Autoclavable | |
Monochrome | |
256 MB | |
76 mm/s | |
Brady™ Workstation Basic Design Suite | |
9.1 kg | |
Component and Equipment Labeling, General Identification, Hazard Identification, Temperature Labeling | |
AC Adaptor |
Thermal Transfer | |
305 mm | |
241 mm | |
260 mm | |
Non-sterile | |
Bulgarian, French, German, Croatian, Italian, Danish, Dutch, Polish, Estonian, Finnish, Romanian, Chinese (Simplified), Slovakian, Slovenian, Spanish, Swedish, Turkish, Chinese (Tradditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish | |
S3100 | |
300 dpi | |
Ethernet, USB | |
UL, cUL, CE, WEEE, RoHS, RCM, EAC, CCC, RoHS 2 | |
1 Year | |
110/240 V |